Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylnaphth[2,3-d]oxazole 97.0+%, TCI America™
SDP

Catalog No. M128910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M128910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M128910G Supplier TCI America Supplier No. M128910G

Chemical Identifiers

CAS 20686-66-2
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00624597
InChI Key WMIHEWJRNFUVNR-UHFFFAOYSA-N
Synonym 2-methylnaphth 2,3-d oxazole, 2-methylnaphtho 2,3-d oxazole, 2-methylbenzo f 1,3 benzoxazole, 2-methyl-naphtho 2,3-d oxazole, 2-methylnaphtho 2,3-d 1,3 oxazole, 2-methylbenzo f benzoxazole, pubchem8702
PubChem CID 140743
IUPAC Name 2-methylbenzo[f][1,3]benzoxazole
SMILES CC1=NC2=CC3=CC=CC=C3C=C2O1

Specifications

Melting Point 87°C
Boiling Point 310°C
Quantity 10 g
Formula Weight 183.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylnaphth[2,3-d]oxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.