Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylnaphthalene 99%, Thermo Scientific™

Catalog No. AC414555000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC414555000 500 g
AC414551000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414555000 Supplier Thermo Scientific Chemicals Supplier No. 414555000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-57-6
Molecular Formula C11H10
Molecular Weight (g/mol) 142.201
InChI Key QIMMUPPBPVKWKM-UHFFFAOYSA-N
Synonym naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10
PubChem CID 7055
ChEBI CHEBI:50720
IUPAC Name 2-methylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 32°C to 35°C
Color White
Density 1
Infrared Spectrum Authentic
Linear Formula C10H7CH3
Quantity 500 g
Beilstein 05, 567
Formula Weight 142.2
Percent Purity ≥98.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Methylnaphthalene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 202-078-3
RTECSNumber QJ9635000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.