Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyloctane 98.0+%, TCI America™
SDP

Catalog No. M03085ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03085ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03085ML Supplier TCI America Supplier No. M03085ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3221-61-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00027307
InChI Key ZUBZATZOEPUUQF-UHFFFAOYSA-N
PubChem CID 18591
IUPAC Name 2-methyloctane
SMILES CCCCCCC(C)C

Specifications

Color Colorless
Boiling Point 143°C
Quantity 5 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyloctane
EINECSNumber (2)-0009
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.