Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpentyl Formate 90.0+%, TCI America™
SDP

Catalog No. F01745ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01745ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01745ML Supplier TCI America Supplier No. F01745ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 381670-34-4
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00059817
InChI Key KYHHSTLLLQRFHJ-UHFFFAOYNA-N
Synonym Formic Acid 2-Methylpentyl Ester
PubChem CID 537217
IUPAC Name 2-methylpentyl formate
SMILES CCCC(C)COC=O

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 mL
UN Number 3272
Formula Weight 130.19
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylpentyl Formate
EINECSNumber (2)-3050
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.