Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpiperazine, 98%, Thermo Scientific™

Catalog No. AA4278630
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4278630 250 g
AA4278618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4278630 Supplier Thermo Scientific Chemicals Supplier No. 4278630

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-07-9
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005954
InChI Key JOMNTHCQHJPVAZ-UHFFFAOYSA-N
Synonym piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
PubChem CID 66057
IUPAC Name 2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 65°C to 67°C
Boiling Point 155°C (763 mmHg)
Flash Point 65°C (149°F)
Quantity 250 g
UN Number UN1325
Formula Weight 100.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Methylpiperazine
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H314-H318-H228
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.