Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpiperidine, 99%, Thermo Scientific™

Catalog No. p-7033153
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5018103 1 g
AAH5018106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5018103 Supplier Thermo Scientific Chemicals Supplier No. H5018103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-05-7
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00005982
InChI Key NNWUEBIEOFQMSS-UHFFFAOYNA-N
Synonym 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline
PubChem CID 7974
IUPAC Name 2-methylpiperidine
SMILES CC1CCCCN1

Specifications

Melting Point -4°C
Boiling Point 118°C to 119°C
Flash Point 8°C (46°F)
Quantity 1 g
UN Number UN2733
Beilstein 79804
Formula Weight 99.18
Percent Purity 99%
Chemical Name or Material 2-Methylpiperidine
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
RTECSNumber TN1240000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.