Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylpyrazine | 109-08-0 | 100.0% | 94.11 g/mol | C5H6N2 | 1 ML
SDP

Catalog No. 5000210392
Encompass_Preferred

2-Methylpyrazine is an alkylpyrazine found in roasted red pepper seed oils. Offered as a high-purity reagent for research and analytical use, it is available as a 10 mM solution in DMSO and in multiple bulk solid sizes.

  • Purity: 99.99% (manufacturer listed)
  • Format: 10 mM solution in DMSO, and solid packs from 10 g to 1 kg
  • Chemical formula: C5H6N2
  • Molecular weight: 94.11 g/mol
  • Storage: store solution and solids at recommended temperatures; short-term shipping at room temperature is acceptable

Catalog No. 50-002-10392 Supplier Medchemexpress LLC Supplier No. HYW06735810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylpyrazine is an alkylpyrazine found in roasted red pepper seed oils. Offered as a high-purity reagent for research and analytical use, it is available as a 10 mM solution in DMSO and in multiple bulk solid sizes.

  • Purity: 99.99% (manufacturer listed)
  • Format: 10 mM solution in DMSO, and solid packs from 10 g to 1 kg
  • Chemical formula: C5H6N2
  • Molecular weight: 94.11 g/mol
  • Storage: store solution and solids at recommended temperatures; short-term shipping at room temperature is acceptable

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.