Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylpyrazine | 109-08-0 | 100.0% | 94.11 g/mol | C5H6N2 | 100 G
SDP

Catalog No. 5000210390
Encompass_Preferred

2-Methylpyrazine (CAS 109-08-0) is an alkylpyrazine used as a research-grade chemical standard and reagent. It has molecular formula C5H6N2 and molecular weight 94.11 g/mol. Typical applications include analytical reference standards and flavor compound studies. Refer to the supplier datasheet and SDS for handling, purity, and storage instructions.

  • High purity (99.99% by GC)
  • Molecular formula C5H6N2
  • Molecular weight 94.11 g/mol
  • Available as solid and solution formats, including 100 G pack
  • Storage: room temperature under nitrogen; in solvent: -80°C (2 years), -20°C (1 year)
  • For research use only; not for human or clinical use

Catalog No. 50-002-10390 Supplier Medchemexpress LLC Supplier No. HYW067358100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Methylpyrazine (CAS 109-08-0) is an alkylpyrazine used as a research-grade chemical standard and reagent. It has molecular formula C5H6N2 and molecular weight 94.11 g/mol. Typical applications include analytical reference standards and flavor compound studies. Refer to the supplier datasheet and SDS for handling, purity, and storage instructions.

  • High purity (99.99% by GC)
  • Molecular formula C5H6N2
  • Molecular weight 94.11 g/mol
  • Available as solid and solution formats, including 100 G pack
  • Storage: room temperature under nitrogen; in solvent: -80°C (2 years), -20°C (1 year)
  • For research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.