Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylpyrazine | 109-08-0 | MFCD00006142 | 100.0% | 94.11 g/mol | C5H6N2 | 25 G
SDP

Catalog No. 5000209512
Encompass_Preferred

2-Methylpyrazine is an alkylpyrazine research chemical found in roasted red pepper seed oils, provided as a high-purity analytical reagent for research and analytical applications.

  • High purity: 99.99%.
  • Cas number: 109-08-0.
  • Molecular formula: C5H6N2.
  • Molecular weight: 94.11 g/mol.
  • Available quantity: 25 G.
  • Storage: room temperature under nitrogen; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Solubility: soluble in DMSO; solution forms available (e.g., 10 mM in DMSO).

Catalog No. 50-002-09512 Supplier Medchemexpress LLC Supplier No. 25GHYW067358
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Methylpyrazine is an alkylpyrazine research chemical found in roasted red pepper seed oils, provided as a high-purity analytical reagent for research and analytical applications.

  • High purity: 99.99%.
  • Cas number: 109-08-0.
  • Molecular formula: C5H6N2.
  • Molecular weight: 94.11 g/mol.
  • Available quantity: 25 G.
  • Storage: room temperature under nitrogen; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Solubility: soluble in DMSO; solution forms available (e.g., 10 mM in DMSO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.