Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methylpyrrolidine

Catalog No. 4780592G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-805-92G 2 g
47-805-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-805-92G Supplier Sigma Aldrich Supplier No. 4780592G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-methylpyrrolidine is a substituted pyrrolidine
  • It is one of the major bioactive compounds found in teucrium manghuaense
  • The hydrodenitrogenation of 2-methylpyrrolidine in the presence of sulfided nimo/α-al2o3 catalyst has been studied.2-methylpyrrolidine may be used as a guest molecule for the synthesis of nonasil-[4158], a clatharate compound

Specifications

CAS 765-38-8
Density 0.834 g/mL (at 25°C (literature))
Boiling Point 97°C to 99°C (744 mmHg)
Molecular Formula C5H11N
Linear Formula C5H11N
Refractive Index n20/D 1.437 (literature)
MDL Number MFCD00014491
Quantity 2 g
Molecular Weight (g/mol) 85.15
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.