Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylquinoline N-Oxide 98.0+%, TCI America™
SDP

Catalog No. M30001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30001G 1 g
M30005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30001G Supplier TCI America Supplier No. M30001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1076-28-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
InChI Key FZJUONUBFWNHNU-UHFFFAOYSA-N
PubChem CID 97325
IUPAC Name 2-methyl-1-oxidoquinolin-1-ium
SMILES CC1=[N+](C2=CC=CC=C2C=C1)[O-]

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylquinoline N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.