Learn More
Sigma Aldrich 2-(Methylsulfanyl)-6-propyl-4(3H)-pyrimidinone
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2-butyl-1-octanol (buoa) is a long-chain glass forming monohydroxy alcohol.2-butyl-1-octanol (buoa) has been used to synthesize:
- 2-butyl-1-octyl-methacrylate (boma)
- 3,5,5-trimethyl-1-hexyl methacrylate (tmhma)
- Hydrophobic polyesters in miniemulsion in the presence of large amounts of waterit has also been used as an extraction solvent in extractive fed-batch experiments
Specifications
Specifications
| CAS | 5751-17-7 |
| Molecular Formula | C8H12N2OS |
| Linear Formula | C8H12N2OS |
| MDL Number | MFCD00847268 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 184.26 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.