Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate | 1280210-80-1 | 98.8% | 10 G
SDP

Supplier:  Medchemexpress LLC HYB078410G

Encompass_Preferred

2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate is a drug intermediate for the synthesis of various active compounds.

  • Target drug intermediate
  • Molecular formula: C12H15N3O5S2
  • Molecular weight: 345.40
  • Appearance: White to light yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Purity: 98.8%

Catalog No. 50-004-08041


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.