Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate | 1280210-80-1 | 98.8% | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYB0784

Encompass_Preferred

2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate is a chemical compound primarily used as an intermediate in drug synthesis. It serves as a versatile building block for creating various active compounds in pharmaceutical research.

  • Functions as a drug intermediate
  • Enables synthesis of various active compounds
  • Purity of 98.84%
  • Molecular formula: C12H15N3O5S2
  • Molecular weight: 345.40

Catalog No. 50-004-05752


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.