Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate | 1280210-80-1 | 98.8% | 5 G
SDP

Supplier:  Medchemexpress LLC HYB07845G

Encompass_Preferred

2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate is a drug intermediate for synthesis of various active compounds. This product has a purity of 98.84%.

  • For research use only
  • Store as powder at -20°C for 3 years
  • Store as powder at 4°C for 2 years
  • Store in solvent at -80°C for 6 months
  • Store in solvent at -20°C for 1 month
  • Appearance: white to light yellow solid

Catalog No. 50-004-08098


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.