Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate | 1280210-80-1 | MFCD28144046 | 98.8% | 1 G
SDP

Supplier:  Medchemexpress LLC HYB07841G

Encompass_Preferred

This product is a drug intermediate, crucial for the synthesis of various active compounds. Its unique chemical structure makes it a versatile building block in pharmaceutical research and development.

  • High purity: 98.8%
  • Molecular formula: C12H15N3O5S2
  • Molecular weight: 345.40
  • Ideal for use as a drug intermediate
  • Stable for long-term storage as powder: -20°C for 3 years, 4°C for 2 years
  • Stable for shorter-term storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-09493


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.