Learn More
Medchemexpress LLC 2-(methylsulfonyl)ethyl n-succinimidyl carbonate | 57903-15-8 | MFCD00043142 | 97.0% | 265.24 g/mol | C8H11NO7S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW319374100MG
Encompass_Preferred
2-(Methylsulfonyl)ethyl N-succinimidyl carbonate is an N-hydroxysuccinimide (NHS) carbonate reagent used for selective modification and crosslinking of primary amines on proteins and peptides. It provides an amino-protecting functionality and a methylsulfonyl-ethyl spacer to enable controlled bioconjugation and analytical workflows.
  • High-purity solid suitable for biochemical reactions.
  • Selective reaction with primary amines via NHS carbonate chemistry.
  • Provides amino-protecting functionality for controlled conjugation.
  • Soluble in common organic solvents for reaction flexibility.
  • Useful for protein crosslinking and bioconjugation method development.
Catalog No. 50-002-80788