Learn More
Medchemexpress LLC 2-Methyltetrahydrofuran-3-one | 3188-00-9 | 99.7% | 100.12 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW010608100G
Encompass_Preferred
2-Methyltetrahydrofuran-3-one is a quorum sensing inhibitor that can inhibit biofilm formation by *H. alvei*. It is also identified as one of the volatile constituents found in roasted coffee.
  • Quorum sensing inhibitor
  • Inhibits biofilm formation
  • Volatile constituent of roasted coffee
  • For research use only
Catalog No. 50-004-53602