Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, 99%, stab. with ca 150-400ppm BHT

Catalog No. AAL073560F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL073560F 2500 mL
AAL07356AE 100 mL
AAL07356AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL073560F Supplier Thermo Scientific Chemicals Supplier No. L073560F
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 96-47-9 | C5H10O | 86.134 g/mol

2-Methyltetrahydrofuran acts as a solvent in organic synthesis. It is considered as a replacement for terahydrofuran due to its higher reaction temperature and easy separation after reaction. It is also useful in the electrolyte formulation for secondary lithium electrodes and as a component in alternative fuels. Further, it is used as a solvent for spectroscopic studies at -1960C. It also acts as a solvent for Grignard reagent in organometallic reactions. In addition to this, it plays an important role as a motor fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyltetrahydrofuran acts as a solvent in organic synthesis. It is considered as a replacement for terahydrofuran due to its higher reaction temperature and easy separation after reaction. It is also useful in the electrolyte formulation for secondary lithium electrodes and as a component in alternative fuels. Further, it is used as a solvent for spectroscopic studies at -1960C. It also acts as a solvent for Grignard reagent in organometallic reactions. In addition to this, it plays an important role as a motor fuel.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point -136°C
Density 0.856
Quantity 2500 mL
Refractive Index 1.406
UN Number UN2536
Beilstein 102448
Boiling Point 80°C to 82°C
Solubility Information Miscible with water and most organic solvents.
Formula Weight 86.13
Percent Purity 99%
Chemical Name or Material 2-Methyltetrahydrofuran, stabilized with ≈150 to 400ppm BHT
Show More Show Less
Hazard Category H225-H302-H315-H318-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLTETRAHYDROFURAN
EINECSNumber 202-507-4
RTECSNumber LU2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.