Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, ReagentPlus™, ≥99.5%,Solstice

Catalog No. 60044900 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
4 x 4 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-900 4 L Glass Bottle
60-044-898 2.5 L Glass Bottle
60-044-901 4 x 4 L Glass Bottle
60-044-897 100 mL Glass Bottle
60-044-902 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-044-900 Supplier Solstice Supplier No. 1558104L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, contains 150-400 ppm BHT as stabilizer,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point <-20°C
Color Colorless
Density 0.86g/mL at 25°C
Quantity 4 L
Flash Point −10°C (14°F)
Packaging Glass Bottle
Refractive Index n20/D 1.406(lit.)
UN Number UN2536
Beilstein 102448
Boiling Point 78°C to 80°C
Formula Weight 86.13g/mol
Percent Purity ≥99.5%
Grade ReagentPlus
Chemical Name or Material 2-Methyltetrahydrofuran
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H302-H315-H318
DOTInformation 3/PGII
EINECSNumber 202-507-4
RTECSNumber LU2800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.