Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, Spectrum™ Chemical
SDP

Catalog No. 18600117 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-117 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-600-117 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M24061LT

M2406, 96-47-9, C5H10O

Spectrum™ Chemical 2-Methyltetrahydrofuran

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key JWUJQDFVADABEY-UHFFFAOYNA-N
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

CAS Max % 100%
Quantity 1 L
Packaging Amber Glass Bottle
UN Number UN2536
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.