Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane) 98.0+%, TCI America™
SDP

Catalog No. M2609200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2609200MG 200 mg
M26091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2609200MG Supplier TCI America Supplier No. M2609200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 892505-41-8
Molecular Formula C10H9BF2N2S
Molecular Weight (g/mol) 238.063
InChI Key OSUYHKLOJHHXAI-UHFFFAOYSA-N
PubChem CID 23631051
SMILES [B-]1(N2C=CC=C2C(=C3[N+]1=CC=C3)SC)(F)F

Specifications

Melting Point 110°C
Color Red
Quantity 200 mg
Formula Weight 238.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material [2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.