Learn More
Medchemexpress LLC 2-Methylthio-AMP | 22140-20-1 | 10MM 1ML
SDP
Catalog No. 5000238205
Encompass_Preferred
2-Methylthio-AMP (2-MeSAMP) is a selective and direct P2Y12 antagonist 2-Methylthio-AMP is an inhibitor of ADP-dependent platelet aggregation[1][2][3]
Catalog No. 50-002-38205 Supplier Medchemexpress LLC Supplier No. HY12598910MM 1ML
2-Methylthio-AMP (2-MeSAMP) is a selective and direct P2Y12 antagonist 2-Methylthio-AMP is an inhibitor of ADP-dependent platelet aggregation[1][2][3]