Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)phenylboronic acid, 97%, Thermo Scientific™

Catalog No. AC393780010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393780010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC393780010 Supplier Thermo Scientific Chemicals Supplier No. 393780010
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 168618-42-6
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.02
MDL Number MFCD01318165
InChI Key QXBWTYBCNFKURT-UHFFFAOYSA-N
Synonym 2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
PubChem CID 2773543
SMILES CSC1=CC=CC=C1B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 84.0°C to 86.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3SC6H4B(OH)2
Quantity 1 g
IUPAC Name (2-methylsulfanylphenyl)boronic acid
Formula Weight 168.02
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)phenylboronic acid
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.