Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M15705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1570-5G 5 g
M15701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1570-5G Supplier TCI America Supplier No. M15705G
Add to Cart
Add to Cart

Specifications

CAS 168618-42-6
Melting Point 83°C
Molecular Formula C7H9BO2S
Quantity 5 g
MDL Number MFCD01318165
Synonym 2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid
InChI Key QXBWTYBCNFKURT-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1SC)(O)O
IUPAC Name (2-methylsulfanylphenyl)boronic acid
Molecular Weight (g/mol) 168.017
PubChem CID 2773543
Formula Weight 168.02
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.