Learn More
Medchemexpress LLC 2-methylthioadenosine diphosphate trisodium | 475193-31-8 | 99.4% | 539.24 | 1 MG
SDP
Catalog No. 5000346658
Encompass_Preferred
2-Methylthioadenosine diphosphate trisodium is a potent purinergic P2Y receptor agonist, with EC50s of 19, 6.2, and 5 nM for human P2Y13, mouse P2Y13 and human P2Y12, respectively. It has pEC50s of 8.29 and 5.75 for human P2Y1 and rat P2Y6, respectively. It induces platelet aggregation and shape change, and inhibits cyclic AMP accumulation in platelets exposed to prostaglandin E1.
  • Potent purinergic P2Y receptor agonist
  • Acts on human P2Y13, mouse P2Y13, human P2Y12, human P2Y1 and rat P2Y6 receptors
  • Induces platelet aggregation and shape change
  • Inhibits cyclic AMP accumulation in platelets exposed to prostaglandin E1
Catalog No. 50-003-46658 Supplier Medchemexpress LLC Supplier No. HY10864810MG
2-Methylthioadenosine diphosphate trisodium is a potent purinergic P2Y receptor agonist, with EC50s of 19, 6.2, and 5 nM for human P2Y13, mouse P2Y13 and human P2Y12, respectively. It has pEC50s of 8.29 and 5.75 for human P2Y1 and rat P2Y6, respectively. It induces platelet aggregation and shape change, and inhibits cyclic AMP accumulation in platelets exposed to prostaglandin E1.
  • Potent purinergic P2Y receptor agonist
  • Acts on human P2Y13, mouse P2Y13, human P2Y12, human P2Y1 and rat P2Y6 receptors
  • Induces platelet aggregation and shape change
  • Inhibits cyclic AMP accumulation in platelets exposed to prostaglandin E1