Learn More
Medchemexpress LLC 2-Methylthioadenosine diphosphate trisodium | 475193-31-8 | 99.9% | 539.24 | 1 MG
SDP
Catalog No. 5000344521
Encompass_Preferred
2-Methylthioadenosine diphosphate trisodium is a potent purinergic P2Y receptors agonist. It induces platelet aggregation and shape change, and inhibits cyclic AMP accumulation in platelets exposed to prostaglandin E1.
  • Potent purinergic P2Y receptors agonist
  • Induces platelet aggregation and shape change
  • Inhibits cyclic AMP accumulation
Catalog No. 50-003-44521 Supplier Medchemexpress LLC Supplier No. HY1086481MG
2-Methylthioadenosine diphosphate trisodium is a potent purinergic P2Y receptors agonist. It induces platelet aggregation and shape change, and inhibits cyclic AMP accumulation in platelets exposed to prostaglandin E1.
  • Potent purinergic P2Y receptors agonist
  • Induces platelet aggregation and shape change
  • Inhibits cyclic AMP accumulation