Learn More
Medchemexpress LLC 2-Methylthiophene | 554-14-3 | 99.9% | 98.16 | 1 G
SDP
Catalog No. 5000453604
Encompass_Preferred
2-Methylthiophene is an intermediate utilized in the synthesis of aromatic sulfur compounds. It appears as a colorless to light yellow liquid with a density of 1.014 g/cm³.
  • Used in the synthesis of aromatic sulfur compounds.
  • Detailed solubility information is provided for in vitro and in vivo applications.
  • Documentation such as data sheet, COA, SDS, HNMR, GC, and handling instructions are available.
  • Storage recommendations are provided for various conditions.
  • Relevant calculators for molarity and dilution are provided.
Catalog No. 50-004-53604 Supplier Medchemexpress LLC Supplier No. HYW0106151G
2-Methylthiophene is an intermediate utilized in the synthesis of aromatic sulfur compounds. It appears as a colorless to light yellow liquid with a density of 1.014 g/cm³.
  • Used in the synthesis of aromatic sulfur compounds.
  • Detailed solubility information is provided for in vitro and in vivo applications.
  • Documentation such as data sheet, COA, SDS, HNMR, GC, and handling instructions are available.
  • Storage recommendations are provided for various conditions.
  • Relevant calculators for molarity and dilution are provided.