Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methylthiophene | 554-14-3 | 99.94% | 98.16 | 5 G
SDP

Catalog No. 5000452924
Encompass_Preferred

2-Methylthiophene is an intermediate used in the synthesis of aromatic sulfur compounds. It is for research use only and not sold to patients. It appears as a colorless to light yellow liquid.

  • Purity: 99.94%
  • Available in various sizes
  • Documentation includes Data Sheet, COA, SDS, and Handling Instructions
  • Soluble in DMSO for in vitro use
  • Storage at 4°C under nitrogen

Catalog No. 50-004-52924 Supplier Medchemexpress LLC Supplier No. HYW0106155G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylthiophene is an intermediate used in the synthesis of aromatic sulfur compounds. It is for research use only and not sold to patients. It appears as a colorless to light yellow liquid.

  • Purity: 99.94%
  • Available in various sizes
  • Documentation includes Data Sheet, COA, SDS, and Handling Instructions
  • Soluble in DMSO for in vitro use
  • Storage at 4°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.