Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylthiophene, 98%

Catalog No. p-3758483
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127850250 25 g Glass bottle
AC127851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC127850250 Supplier Thermo Scientific Chemicals Supplier No. 127850250
Only null left
Add to Cart

CAS: 554-14-3 | C5H6S | 98.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-14-3
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.16
MDL Number MFCD00005451
InChI Key XQQBUAPQHNYYRS-UHFFFAOYSA-N
Synonym thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
PubChem CID 11126
IUPAC Name 2-methylthiophene
SMILES CC1=CC=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -63°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 113°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5190 to 1.521
Quantity 25 g
Beilstein 17, 37
Specific Gravity 1.02
Solubility Information Solubility in water: immiscible
Formula Weight 98.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylthiophene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-063-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.