Learn More
Description
- 2-chloro-n,n-diethylacetamide has been used in:
- Preparation of calixarene amides
- Synthesis of 5-bromo-25,26,27,28-tetrakis[(n,n-diethylaminocarbonyl)-methoxy]calix[4]arene
- Synthesis of amide calix[4]pyrrole macrocycles with super-extended cavities
- Microwave-assisted organic synthesis of 3-cyano-n,n-diethyl-4-(4-methoxyphenoyl)-4-oxobutanamide
Specifications
Specifications
| CAS | 823-09-6 |
| Molecular Formula | C5H6N2S |
| Linear Formula | C5H6N2S |
| MDL Number | MFCD09025802 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 126.18 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.