Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylundecanal 95%, Thermo Scientific™

Catalog No. AC127980250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC127980250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC127980250 Supplier Thermo Scientific Chemicals Supplier No. 127980250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-41-8
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
InChI Key NFAVNWJJYQAGNB-UHFFFAOYSA-N
Synonym undecanal, 2-methyl, aldehyde c-12, aldehyde m.n.a., methyl-n-nonylacetaldehyde, methylnonylacetaldehyde, 2-methylhendecanal, methylnonylacetic aldehyde, aldehyde c-12 mna, 2-methyl undecanal, methyl nonyl acetic aldehyde
PubChem CID 61031
IUPAC Name 2-methylundecanal
SMILES CCCCCCCCCC(C)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.829
Infrared Spectrum Authentic
Refractive Index 1.4300 to 1.4350 (20°C, 589nm)
Linear Formula CH3(CH2)8CH(CH3)CHO
Quantity 25 mL
Specific Gravity 0.824 to 0.834 (25°/25°C)
Formula Weight 184.32
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 2-Methylundecanal, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.