Learn More
Medchemexpress LLC 2-Methylvaleric acid | 97-61-0 | 98.0% | 116.16 | 50 G
SDP
Catalog No. 5000452918
Encompass_Preferred
2-Methylvaleric acid is a branched short-chain fatty acid (SCFA) produced by the metabolism of branched-chain amino acids by gut microbes. It can serve as a potential biomarker for metabolic diseases like type 2 diabetes, where its content is significantly reduced in the feces of diabetic mice. This compound may regulate host energy metabolism and inflammatory response through G protein-coupled receptors (GPCRs) or histone deacetylase (HDAC) inhibition. It is suitable for studying gut microbe-host interactions and metabolic diseases as a fecal biomarker.
  • Liquid appearance with a density of 0.931 g/cm³
  • Colorless to light yellow
  • Derived from plants or as an endogenous metabolite
  • Classified as ketones, aldehydes, and acids
  • Suitable for studying gut microbe-host interactions and metabolic diseases
  • For research use only
Catalog No. 50-004-52918 Supplier Medchemexpress LLC Supplier No. HYW01051650G
2-Methylvaleric acid is a branched short-chain fatty acid (SCFA) produced by the metabolism of branched-chain amino acids by gut microbes. It can serve as a potential biomarker for metabolic diseases like type 2 diabetes, where its content is significantly reduced in the feces of diabetic mice. This compound may regulate host energy metabolism and inflammatory response through G protein-coupled receptors (GPCRs) or histone deacetylase (HDAC) inhibition. It is suitable for studying gut microbe-host interactions and metabolic diseases as a fecal biomarker.
  • Liquid appearance with a density of 0.931 g/cm³
  • Colorless to light yellow
  • Derived from plants or as an endogenous metabolite
  • Classified as ketones, aldehydes, and acids
  • Suitable for studying gut microbe-host interactions and metabolic diseases
  • For research use only