Learn More
Cayman Chemical 2-MIcroglobulIndog Moncl 100ug
SDP
Catalog No. 502948978
Encompass_Preferred
Immunogen: Recombinant dog β2-microglobulin protein (amino acids 28-125) • Host: Mouse • Species Reactivity: (+) Dog • Cross Reactivity: (+) β2-microglobulin • Applications: FC, IP • MW = 14 kDa
Catalog No. 50-294-8978 Supplier Cayman Chemical Supplier No. 33370100 UG
Immunogen: Recombinant dog β2-microglobulin protein (amino acids 28-125) • Host: Mouse • Species Reactivity: (+) Dog • Cross Reactivity: (+) β2-microglobulin • Applications: FC, IP • MW = 14 kDa