Learn More
OHAUS 2 Microplate Foam Insert
Catalog No. 01804388 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-388 Vortex Attachment
1 options
Catalog No. 01-804-388 Supplier OHAUS Supplier No. 83041417
Add to Cart
Add to Cart
Two high-density foam inserts to accommodate one standard footprint microplate.
  • This insert fits into the 6 in. (152 mm) Platform #146-6005-00 attachment for use with the Vortex-Genie 2 mixers and Vortex-Genie Pulse.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Microplate Insert for Recessed Platform Head