Learn More
Alkali Scientific Alkali Scientific™ 2" Mini White Cardboard Freezer Box, 16 Cell Divider
SDP
Catalog No. 503026884
Encompass_Preferred
Alkali Scientific™ 2" mini cardboard freezer box are ideal for any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cryo-freezer boxes are protected from liquid nitrogen or condensation from freezers by a moisture repellent coating.  Our freezer boxes are made of a sturdy white cardbard and fit all standard-stainless steel and polycarbonate storage systems.  Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C. 
Catalog No. 50-302-6884 Supplier Alkali Scientific Supplier No. MB2C16
Alkali Scientific™ 2" mini cardboard freezer box are ideal for any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cryo-freezer boxes are protected from liquid nitrogen or condensation from freezers by a moisture repellent coating.  Our freezer boxes are made of a sturdy white cardbard and fit all standard-stainless steel and polycarbonate storage systems.  Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C. 