Learn More
Perkin Elmer US LLC 2 mL Amber Glass 9 mm Screw Top Vial with Write-On Patch, 100/pk
Catalog No. 502103702
Encompass_Preferred
Our vials are manufactured from Type I Borosilicate Glass, which meets all USP, JP, and EP Pharmacopeia requirements. The glass performs excellent at high temperatures and is chemical resistant to acidic, neutral and alkali solutions. All our vials are packed in a clean environment to ensure you receive contaminant free product every time.
Catalog No. 50-210-3702 Supplier Perkin Elmer US LLC Supplier No. N9307802
Our vials are manufactured from Type I Borosilicate Glass, which meets all USP, JP, and EP Pharmacopeia requirements. The glass performs excellent at high temperatures and is chemical resistant to acidic, neutral and alkali solutions. All our vials are packed in a clean environment to ensure you receive contaminant free product every time.