Learn More
Perkin Elmer US LLC 2 mL Autosampler Convenience Kit - 10 mm Screw Top Amber Glass Vial with PTFE/Silicone Pre-Slit Septa, 100/pk
Catalog No. 502093376
Encompass_Preferred
Vial Top Type Vial Volume Capacity Vial Material Neck I.D. Size Cap Type Septa Material Septa Type Quantity Screw Top 2 mL Amber Glass 10 mm Black Cap PTFE/Silicone Pre-Slit 100
Catalog No. 50-209-3376 Supplier Perkin Elmer US LLC Supplier No. N9300696
Vial Top Type Vial Volume Capacity Vial Material Neck I.D. Size Cap Type Septa Material Septa Type Quantity Screw Top 2 mL Amber Glass 10 mm Black Cap PTFE/Silicone Pre-Slit 100