Learn More
Waters Corp HPLC Vial Rack, 2 mL, Domino
Catalog No. NC3762195 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 2 mL HPLC Vial Rack Domino is designed to hold in place a variety of racks commonly used to hold HPLC vials in various formats. Dominos are machine-readable consumable holders that create the interface between a variety of different consumable types and the Andrew+ pipetting robot to enable seamless automation of many different laboratory workflows.
  • Equipped with QR codes housing critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
  • Indispensable part of any Andrew+ automation configuration.
  • Designed to hold various HPLC vial racks.
  • Facilitates seamless automation of laboratory workflows with the Andrew+ pipetting robot.
Catalog No. NC3762195 Supplier Waters Corp Supplier No. 186010091
The 2 mL HPLC Vial Rack Domino is designed to hold in place a variety of racks commonly used to hold HPLC vials in various formats. Dominos are machine-readable consumable holders that create the interface between a variety of different consumable types and the Andrew+ pipetting robot to enable seamless automation of many different laboratory workflows.
  • Equipped with QR codes housing critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
  • Indispensable part of any Andrew+ automation configuration.
  • Designed to hold various HPLC vial racks.
  • Facilitates seamless automation of laboratory workflows with the Andrew+ pipetting robot.