Learn More
Perkin Elmer US LLC 2 mL Pre-Assembled 9 mm Blue Polypropylene Screw Cap with Ultra-Low Bleed PTFE/Silicone Pre-Slit Septa, 100/pk
Supplier:  Perkin Elmer US LLC N9306364
Encompass_Preferred
Cap Type Cap Material Neck I.D. Size: Septa Material Septa Features Quantity Screw Top Blue Polypropylene 9 mm PTFE/Silicone Ultra-Low Bleed, Pre-Slit 100
Catalog No. 50-211-3661