Learn More
Chemglass Life Sciences Stopcock, 120° Bore, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG44001
Encompass_Preferred
Stopcock, 120¿ Bore, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE stopcock with three sidearms sealed 120¿ apart. Plug has a bore to allow connection of any two arms at once.
Catalog No. CG44001 Supplier Chemglass Life Sciences Supplier No. CG44001
Stopcock, 120¿ Bore, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE stopcock with three sidearms sealed 120¿ apart. Plug has a bore to allow connection of any two arms at once.