Learn More
World Precision Instrument 2 mm Receptacle to 0.031" Jack (for Omega-TipZ)
SDP
Catalog No. 502521250
Encompass
Jack for the Omega-TipZ impedance measurement system
You can use this adapter to connect the metal microelectrodes to the probe. It has a 2mm receptacle  on one end and a 0.031" jack. This is designed for use with the Omega-TipZ battery operated impedance measurement system.
Catalog No. 50-252-1250 Supplier World Precision Instrument Supplier No. 5468
Jack for the Omega-TipZ impedance measurement system
You can use this adapter to connect the metal microelectrodes to the probe. It has a 2mm receptacle  on one end and a 0.031" jack. This is designed for use with the Omega-TipZ battery operated impedance measurement system.