Learn More
Abcam 2 mm Sagittal Mouse Brain Slicer, 1UNIT
Catalog No. 502675570
Encompass_Preferred
This 2 mm Sagittal mouse brain slicer (ab243313) allows you to easily cut a brain into smaller pieces for easier imaging with objectives that have a limited working distance.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-5570 Supplier Abcam Supplier No. AB2433131UNIT
This 2 mm Sagittal mouse brain slicer (ab243313) allows you to easily cut a brain into smaller pieces for easier imaging with objectives that have a limited working distance.
The product is subject to the following: Abcam Restricted Use Statement