Learn More
Perkin Elmer US LLC 2 mm, Splitless Siltek Liner with Wool
Catalog No. 502104582
Encompass_Preferred
Siltek Deactivated Liner (with wool) for Splitless Operation - Optimum sample dispertion for active samples. Surface provides inertness over a wide sample pH range. Wool can beadsorptive if adsorptive if fibers are broken. Quartz Liner for Split Operation.
Catalog No. 50-210-4582 Supplier Perkin Elmer US LLC Supplier No. N6502004
Siltek Deactivated Liner (with wool) for Splitless Operation - Optimum sample dispertion for active samples. Surface provides inertness over a wide sample pH range. Wool can beadsorptive if adsorptive if fibers are broken. Quartz Liner for Split Operation.