Learn More
Cambridge Isotope Laboratories 2 mM Sucrose 0 5 mM DSS 2 mM NaN3 in 90% H2O/10% D2O H2O D2O v/v 1 5mm tube
Catalog No. 5000166567
Encompass
2 mM Sucrose 0 5 mM DSS 2 mM NaN3 in 90% H2O/10% D2O H2O D2O v/v 1 5mm tube
Catalog No. 50-001-66567 Supplier Cambridge Isotope Laboratories Supplier No. DLM91
2 mM Sucrose 0 5 mM DSS 2 mM NaN3 in 90% H2O/10% D2O H2O D2O v/v 1 5mm tube