Learn More
Perkin Elmer US LLC 2 mm, Ultra Deactivated Straight Splitless Liner with Wool
Catalog No. 502111558
Encompass_Preferred
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples. Inlet liners for splitless injection are generally designed as straight tubes, although new designs such as the gooseneck will help contain the sample in the injector. Packing these liners with wool will also help trap non-volatile residue and prevent column contamination. For the capillary injector on the Clarus 590 or 690 GC system.
Catalog No. 50-211-1558 Supplier Perkin Elmer US LLC Supplier No. N6502033
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples. Inlet liners for splitless injection are generally designed as straight tubes, although new designs such as the gooseneck will help contain the sample in the injector. Packing these liners with wool will also help trap non-volatile residue and prevent column contamination. For the capillary injector on the Clarus 590 or 690 GC system.