Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(morpholino)ethylazide

Catalog No. CDS005526 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005526 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005526 Supplier Sigma Aldrich Supplier No. CDS005526100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 2-octynoate has been quantitated as suspected allergen in cosmetic products by headspace-programmed temperature vaporization-fast gas chromatography-quadrupole mass spectrometry
  • It participates in nickel-catalyzed carbocyclization of 2-iodoacetophenone to yield a 2,3-disubstituted indenol

Specifications

Molecular Formula C6H12N4O
Linear Formula C6H12N4O
MDL Number MFCD01320909
Quantity 25 mL
Molecular Weight (g/mol) 156.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.