Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Morpholinoethanesulphonic acid | 4432-31-9 | 554642 | 99.9% | 500 MG
SDP

Catalog No. 5000410119
Encompass_Preferred

MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.

  • Amphoteric ion buffer
  • Buffer capacity: pH 5.5-7.0
  • Widely used in biochemistry and molecular biology experiments
  • Applications include cell culture, enzyme activity determination, electrophoresis, and protein studies

Catalog No. 50-004-10119 Supplier Medchemexpress LLC Supplier No. HYD0858500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.

  • Amphoteric ion buffer
  • Buffer capacity: pH 5.5-7.0
  • Widely used in biochemistry and molecular biology experiments
  • Applications include cell culture, enzyme activity determination, electrophoresis, and protein studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.