Learn More
Medchemexpress LLC 2-Morpholinoethanesulphonic acid hemisodium | 117961-21-4 | 98.0% | 100 G
SDP
Catalog No. 5000409844
Encompass_Preferred
MES (2-Morpholinoethanesulphonic acid) hemisodium is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely utilized in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • It is a zwitterionic buffer and provides effective buffering capacity around pH 6.15.
  • The pKa value changes with temperature, requiring consideration for its use at different temperatures.
  • It shows good buffering properties in biological experiments.
  • It exhibits poor affinity with metal ions, allowing its use in studies involving metal ions.
Catalog No. 50-004-09844 Supplier Medchemexpress LLC Supplier No. HYD0858D100G
MES (2-Morpholinoethanesulphonic acid) hemisodium is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely utilized in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • It is a zwitterionic buffer and provides effective buffering capacity around pH 6.15.
  • The pKa value changes with temperature, requiring consideration for its use at different temperatures.
  • It shows good buffering properties in biological experiments.
  • It exhibits poor affinity with metal ions, allowing its use in studies involving metal ions.