Learn More
Medchemexpress LLC 2-Morpholinoethanesulphonic acid hydrate | 1266615-59-1 | 99.9% | 50 G
SDP
Catalog No. 5000410059
Encompass_Preferred
MES hydrate is an amphoteric ion buffer with a buffering capacity ranging from pH 5.5-7.0. It is widely used in biochemistry and molecular biology experiments for applications such as cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Provides effective buffering capacity at approximately pH 6.15.
  • Its pKa value changes with temperature, necessitating consideration for different experimental conditions.
  • Exhibits good buffering properties in biological experiments.
  • Shows poor affinity with metal ions, allowing its use in studies involving them.
Catalog No. 50-004-10059 Supplier Medchemexpress LLC Supplier No. HYD0858C50G
Add to Cart
Add to Cart
MES hydrate is an amphoteric ion buffer with a buffering capacity ranging from pH 5.5-7.0. It is widely used in biochemistry and molecular biology experiments for applications such as cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Provides effective buffering capacity at approximately pH 6.15.
  • Its pKa value changes with temperature, necessitating consideration for different experimental conditions.
  • Exhibits good buffering properties in biological experiments.
  • Shows poor affinity with metal ions, allowing its use in studies involving them.